Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

182060

Sigma-Aldrich

Poly(2-ethylhexyl acrylate) solution

average Mw ~92,000 by GPC, in toluene

Synonym(s):

2-Ethylhexyl acrylate homopolymer, Poly(ethylhexyl acrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH[CO2CH2CH(C2H5)(CH2)3CH3])n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

1.2 (37 °C, vs air)

mol wt

average Mw ~92,000 by GPC

concentration

in toluene

weight

Unit weight does not include weight of solvent.

transition temp

Tg −55 °C

density

0.903 g/mL at 25 °C

SMILES string

CCCCC(CC)OC(=O)C=C

InChI

1S/C11H20O2/c1-4-7-8-10(5-2)9-13-11(12)6-3/h6,10H,3-5,7-9H2,1-2H3

InChI key

GOXQRTZXKQZDDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(2-ethylhexyl acrylate) solution can be prepared by polymerizing 2-ethylhexyl acrylate. It may also be synthesized by solution polymerization in toluene.

Application

Poly(2-ethylhexyl acrylate) solution may be used as plasticizer for medical grade pressure sensitive adhesives.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal stability of poly (2-ethylhexyl acrylates) used as plasticizers for medical application.
Czech Z, et al.
Polym. Bull., 70(6), 1911-1918 (2013)
The behaviour of poly (2-ethylhexyl acrylate) in dilute solution: viscosity measurements.
Lathova E, et al.
Polym. Bull., 30(6), 713-718 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service