Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

181404

Sigma-Aldrich

Poly(butyl acrylate) solution

average Mw ~99,000 by GPC, in toluene

Synonym(s):

Acrylic acid butyl ester polymer, Butyl acrylate homopolymer, Butyl acrylate polymer, Poly(n -butyl acrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7H12O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

1.2 (37 °C, vs air)

form

liquid

autoignition temp.

997 °F

mol wt

average Mw ~99,000 by GPC

composition

solids, 25-30 wt. %

concentration

in toluene

refractive index

n20/D 1.466 (lit.)

transition temp

Tg -49 °C

density

1.087 g/mL at 25 °C

SMILES string

CCCCOC(=O)C=C

InChI

1S/C7H12O2/c1-3-5-6-9-7(8)4-2/h4H,2-3,5-6H2,1H3

InChI key

CQEYYJKEWSMYFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Unit weight does not include weight of solvent.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service