Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

205591

Sigma-Aldrich

Sodium borodeuteride

98 atom % D, 90% (CP)

Synonym(s):

Deuterated sodium borohydride, Sodium tetradeuterioboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBD4
CAS Number:
Molecular Weight:
41.86
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

90% (CP)

form

powder or chunks

autoignition temp.

752 °F

mp

>300 °C (dec.) (lit.)

density

1.187 g/mL

application(s)

pharmaceutical

mass shift

M+4

SMILES string

[Na+].[2H][B-]([2H])([2H])[2H]

InChI

1S/BH4.Na/h1H4;/q-1;+1/i1D4;

InChI key

YOQDYZUWIQVZSF-XWFVQAFUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium borodeuteride is an inorganic compound with chemical formula NaBD4. The sodium borodeuteride is a stable isotope and is commonly used as a reducing agent.

Application

Sodium borodeuteride is used:
  • As a reducing agent in the asymmetric reduction of carbonyl groups in presence Sn-mediated catalyst.
  • In the deuteration of quinoxalinones with olefins in presence of Iron(III) promoted catalyst.
  • In the synthesis of amine borane and amine trideuterioborane adducts in presence of NaBH4(sodium borohydride) and crown-ether-catalyst.

Features and Benefits

Sodium borodeuteride is
  • An easily handled reducing agent.
  • Commercially available deuteride source.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Organometallic Chemistry, 469, 15-15 (1994)
Synthesis, 4013-4013 (2006)
Tetrahedron Letters, 48, 2335-2335 (2007)
Khalef Lefsih et al.
Phytochemistry, 154, 47-55 (2018-07-15)
This work aims to fill the gap in the present knowledge about the structure of pectin from Opuntia ficus-indica. The water-soluble pectin (WSP) fraction, extracted with the Microwave Assisted Extraction (MAE), was further deproteinated (dWSP) and analyzed through several biophysical
Xiao-Dan Shi et al.
Food chemistry, 271, 338-344 (2018-09-22)
Dictyophora echinovolvata is a kind of edible mushroom in the Dictyophora genus, of which polysaccharide is an important chemical substance. Herein, three polysaccharide fractions (DEP-4P, DEP-6P and DEP-8P) were prepared from water extract of D. echinovolvata using gradient ethanol precipitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service