Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

157333

Sigma-Aldrich

3-Chloroanisole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OCH3
CAS Number:
Molecular Weight:
142.58
Beilstein:
2041497
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.536 (lit.)

bp

193 °C (lit.)

density

1.164 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

COc1cccc(Cl)c1

InChI

1S/C7H7ClO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3

InChI key

YUKILTJWFRTXGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

One-step preparation of 3-chloroanisole from the corresponding 3-substituted nitrobenzene has been reported.

Application

3-Chloroanisole was employed as starting reagent in the regioselective synthesis of 4- and 7-alkoxyindoles. It was also employed as electrolyte additive for the overcharging protection of Li-ion cell.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-Chloroanisole for overcharge protection of a Li-ion cell.
Lee Y-G and Cho J.
Electrochimica Acta, 52(25), 7404-7408 (2007)
Roberto Sanz et al.
The Journal of organic chemistry, 72(14), 5113-5118 (2007-06-15)
An efficient and regioselective synthesis of 4- and 7-alkoxyindoles has been developed from commercially available starting materials such as 3-halophenols and 3-chloroanisole. Directed ortho-metalation followed by two palladium-catalyzed processes, a Sonogashira coupling and a tandem amination/cyclization reaction, allows the synthesis
One-step preparation of some 3-substituted anisoles.
Zilberman J.
Organic Process Research & Development, 7(3), 303-305 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service