Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18116

Sigma-Aldrich

1,8-Diamino-3,6-dioxaoctane

for synthesis

Synonym(s):

1,8-Diamino-3,6-dioxaoctane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16N2O2
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352116
EC Index Number:
213-203-6
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

transition temp

flash point 121 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.011 - 1.014
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service