Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

130265

Sigma-Aldrich

2-Nitropropane

≥96%

Synonym(s):

Isonitropropane, Nitro propane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NO2)CH3
CAS Number:
Molecular Weight:
89.09
Beilstein:
1740684
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~3 (vs air)

Quality Level

vapor pressure

~13 mmHg ( 20 °C)

Assay

≥96%

autoignition temp.

802 °F

refractive index

n20/D 1.394 (lit.)

bp

120 °C (lit.)

mp

−93 °C (lit.)

solubility

H2O: slightly soluble
organic solvents: miscible

density

0.992 g/mL at 25 °C (lit.)

functional group

amine
nitro

SMILES string

CC(C)[N+]([O-])=O

InChI

1S/C3H7NO2/c1-3(2)4(5)6/h3H,1-2H3

InChI key

FGLBSLMDCBOPQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Nitropropane can be converted to mutagenic metabolites by human SULT1A1.

Application

2-Nitropropane was used to prepare 5,5-Dimethylpyrroline N-oxide and 5,5-di(trideuteromethyl)pyrroline N-oxide. It was also used to determine the concordance of results for a pair of structural isomers, 2-nitropropane (2-NP) and 1-nitropropane (1-NP).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 3 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Doi et al.
Journal of toxicologic pathology, 24(4), 207-213 (2012-02-10)
This study was conducted to determine the concordance of results for a pair of structural isomers, 2-nitropropane (2-NP) and 1-nitropropane (1-NP), using the rat medium-term liver carcinogenesis bioassay (Ito test) and previously published long-term carcinogenicity tests. Male F344 rats were
Bente Riis et al.
DNA repair, 1(9), 709-717 (2003-01-02)
This study was set up to investigate the relationships between the formation and removal of DNA damage in form of 8-oxodeoxyguanosine (8-oxodG) in neonatal (day 16 of gestation) as compared to adult rats. The hypothesis addressed was whether the rapidly
Kevin Francis et al.
The Journal of biological chemistry, 280(7), 5195-5204 (2004-12-08)
2-Nitropropane dioxygenase (EC 1.13.11.32) catalyzes the oxidation of nitroalkanes into their corresponding carbonyl compounds and nitrite. In this study, the ncd-2 gene encoding for the enzyme in Neurospora crassa was cloned, expressed in Escherichia coli, and the resulting enzyme was
Reversal of enantioselectivity by tuning the conformational flexibility of phase-transfer catalysts.
Ming-Qing Hua et al.
Angewandte Chemie (International ed. in English), 49(15), 2772-2776 (2010-03-17)
Kirk Simon et al.
Mechanisms of ageing and development, 130(9), 637-647 (2009-08-15)
The p53 DNA damage response attenuated with age and we have evaluated downstream factors in the DNA damage response. In old animals p21 protein accumulates in the whole cell fraction but significantly declines in the nucleus, which may alter cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service