Skip to Content
MilliporeSigma

259489

1-Nitrobutane

95%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)3NO2
CAS Number:
Molecular Weight:
103.12
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
210-980-3
Beilstein/REAXYS Number:
1743017
MDL number:
eCl@ss:
32150312
Assay:
95%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

95%

form

liquid

refractive index

n20/D 1.410 (lit.)

bp

152-153 °C (lit.)

mp

−81 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

functional group

amine, nitro

SMILES string

CCCC[N+]([O-])=O

InChI

1S/C4H9NO2/c1-2-3-4-5(6)7/h2-4H2,1H3

InChI key

NALZTFARIYUCBY-UHFFFAOYSA-N

General description

Low-energy velocity map photoelectron imaging of 1-nitrobutane anions by photoelectron spectroscopy has been reported. Hydrothermal reaction of 1-nitrobutane in high-temperature water has been reported.


Pictograms

Flame

Signal Word

Warning

Hazard Codes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

Flash Point (°F)

111.2 °F - closed cup

Flash Point (°C)

44 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library