Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

109932

Sigma-Aldrich

4-Ethylmorpholine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
102969
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39162412
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.441 (lit.)

bp

139 °C (lit.)

mp

−63 °C (lit.)

density

0.91 g/mL at 20 °C (lit.)

SMILES string

CCN1CCOCC1

InChI

1S/C6H13NO/c1-2-7-3-5-8-6-4-7/h2-6H2,1H3

InChI key

HVCNXQOWACZAFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-ethylmorpholine is used to couple the carboxylic group of lactone in anhydrous DMF.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of 6-carboxynaphthofluorescein phosphoramidite in the automated synthesis of quencher-dye oligonucleotide probes (QDOPs).
Vu, Huynh, et al.
Tetrahedron Letters, 50(7), 737-740 (2009)
R J Rando et al.
American Industrial Hygiene Association journal, 50(1), 8-14 (1989-01-01)
A passive monitor for TDI which utilizes microporous Teflon diffusion membranes and a 0.5% sulfuric acid (H2SO4) absorbing medium was evaluated. The collection rate of TDI is proportional to exposure dose from 10 to at least 300 ppb.hr, and response
V S Kamanna et al.
Biochimica et biophysica acta, 1043(1), 119-121 (1990-03-12)
Apolipoprotein B (ApoB) in general is known as an insoluble protein in aqueous buffers without the initial aid of denaturing agents. However, following the total delipidization of rat plasma low-density lipoproteins, a considerable portion (28.2 +/- 3.0%) of rat ApoB
Seh-Jin Yang et al.
Organic letters, 11(15), 3438-3441 (2009-07-09)
A new, nondestructive, highly sensitive method for colorimetric monitoring of primary amines, secondary amines, and thiols on a solid support was developed. The resin used in this method is simply regenerated for the repetition of the reaction or an ensuing
J-Y Jo et al.
International journal of oral and maxillofacial surgery, 44(7), 921-928 (2015-03-15)
To investigate the effect of the sequential delivery of bone morphogenetic proteins BMP-2 and BMP-7 on bone regeneration in rat calvarial defects (40 Sprague-Dawley rats, 8mm defect size), all animals were treated with a hydroxyapatite (HA)/tricalcium phosphate (TCP) bone graft

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service