Documentos clave
Z757454
Sartorius Proline® Plus multichannel mechanical pippettor
8-channel, volume 10-100 μL
Seleccione un Tamaño
1 EA
US$ 1.090,00
US$ 1.090,00
PRICE WITHOUT NATIONAL TAXES
Fecha estimada de envío31 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 1.090,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 1.090,00
PRICE WITHOUT NATIONAL TAXES
Fecha estimada de envío31 de mayo de 2025
Características
8-channel
electronic: no
envase
pack of 1 ea
fabricante / nombre comercial
Sartorius 728130
volumen
10-100 μL
Categorías relacionadas
Características y beneficios
  • The click-stop system enables easy volume adjustment.
  • Broad volume display for better reading.
  • Color-coded for easy matching of fitting tips
  • High chemical and UV-resistance materials to ensure a long lifetime of the pipette.
  • Fully autoclavable and requires no disassembly.
  • Easy cleaning and maintenance with only three parts to disassemble.
  • Safe-cone filters provided for models >10μL.
  • Εrgοnomic finger support reduces the grip force required to hold the pipette.
  • Range of volume-3μL (5μL for fixed) to 10mL
  • Low pipetting forces
  • Wide range of adjustable single and multi-channel models and fixed single-channel models
Información legal
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)