BAF134730001
Bel-Art® Write-on™ Tape Dispenser
Wall Mount Adapter for Economy Model
About This Item
Código UNSPSC:
55121611
NACRES:
NB.35
Productos recomendados
envase
pack of 1 ea
fabricante / nombre comercial
Bel-Art F13473-0001
L × An
100 mm × 125 mm
Descripción general
Use to mount the dispenser vertically or horizontally.
U-shaped stainless steel foot positions ′Write-On Label Tape Dispenser at a 45° angle off of the wall for improved ergonomics.
U-shaped stainless steel foot positions ′Write-On Label Tape Dispenser at a 45° angle off of the wall for improved ergonomics.
Información legal
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)