Documentos
AXYSCT150OS
Screw cap tubes and caps with O-rings, conical bottom
capacity 1.5 mL, orange, sterile
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
orange
screw top cap
sterility
sterile
feature
RCF 20,000 × g
packaging
pack of 500 tubes
case of 8 packs
manufacturer/tradename
Axygen Inc
Corning
capacity
1.5 mL