Documentos clave
Z678031
Finnpipette® F2 multichannel pipette
volume 5-50 μL
Seleccione un Tamaño
1 EA
US$ 1.700,00
US$ 1.700,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 1.700,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 1.700,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Características
12-channel
color-coded yellow
electronic: no
envase
pack of 1 ea
fabricante / nombre comercial
Finn 4662050
volumen
5-50 μL
Descripción general
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50 μl and below
Información legal
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)