Documentos clave
Z314994
Hirschmann® pipetus®-Standard pipette controller
with pump and sterile filter
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
En este momento no podemos mostrarle ni los precios ni la disponibilidad
entrada CA/CC
230 V AC
Materiales
plastic
silicone
Características
CE compliant
fabricante / nombre comercial
Hirschmann 9904000
disponibilidad
not available in USA and Canada
volumen
0.1-200 mL
Descripción general
Similar to the Pipetus-Akku, but with separate pump unit. The separate pumps avoids any contamination and lowers ignition risk. There are no electronic parts within the controller.
Supplied with tubing, hand-held controller, pipette holder, spare filters, spare safety valve and instruction manual.
UK & European version supplied with 2-pin plug and 3-pin adapter.
Sterile filter mounted on pump, provides clean sterile airflow if required.
Información legal
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Pipetus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)