Merck
Z266051
Supelco
PTFE multi-plate racks
For use with 20 cm × 20 cm plates
About This Item
Código UNSPSC:
41115711
NACRES:
NC.07
técnicas
thin layer chromatography (TLC): suitable
Descripción general
Para utilizar con cubetas de revelado Latch-Lid¿ (de tapa con pestillo). Pueden alojar hasta seis placas de CCF (TLC).
Información legal
Latch-Lid is a trademark of General Glassblowing, Inc.
Producto relacionado
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Christiane Mühle et al.
Journal of chromatography. A, 1481, 137-144 (2016-12-26)
As part of the sphingomyelin pathway, sphingomyelinases and ceramidases have attracted much attention in basic as well as clinical research. However, current assays still often rely on a radioactive substrate, extensive manual purification steps, and hazardous solvents for chromatographic analysis.