Z117145
Stainless steel 304 syringe needle
gauge 22, L 10 in.
About This Item
Código UNSPSC:
41122002
eCl@ss:
32070500
NACRES:
NB.03
Productos recomendados
descripción
Standard Hub with 304 Stainless Steel Tubing
envase
pack of 1 ea
fabricante / nombre comercial
Cadence Science
Longitud
10 in.
manómetro
22
Categorías relacionadas
Descripción general
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Standard Hub with 304 Stainless Steel Tubing
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)