Documentos clave
Z104353
Sigma-Aldrich
Tubing quick-disconnects
for tubing i.d., 1/4 ‑ 5/16 in.
Seleccione un Tamaño
12 EA
US$ 58,80
US$ 58,80
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
12 EA
US$ 58,80
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
US$ 58,80
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
PE
Características
for tubing i.d. 1/4 ‑ 5/16 in.
envase
pack of 12 ea
fabricante / nombre comercial
Bel-Art H19728-0000
Descripción general
Consists of two serrated connectors which are joined securely through a male-female center taper. Connector of one size can be interchanged with other sizes to produce reducing combinations.
  • Easy-to-assemble barbed connectors require no tools
  • Ideal for use with flexible tubing
  • Handy on lines that require frequent disconnection
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)