Documentos clave
Z553379
Sigma-Aldrich
Aldrich® automatic shut-off quick-disconnect
Coupling insert
Seleccione un Tamaño
1 EA
US$ 66,90
US$ 66,90
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 66,90
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
US$ 66,90
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
descripción
GL 14 inner thread
Descripción general
Replace SafetyBarb tubing connectors with these new quick-disconnect fittings, which are made specifically for use with Aldrich glassware. To install quick-disconnects, unscrew tubing connectors on glassware and replace with coupling inserts Z55,337-9. Order one coupling insert and one coupling body for each glass connection. Chemically resistant acetal coupling insert and body are spring-loaded, locking, and have 316SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically. For use with flexible 1/4 in. I.D. tubing.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)