PL800GH-8-10
Plas-Labs Hypalon™ glove box gloves
for 1 piece glove set, size 8 (for 10 in. ports)
About This Item
Código UNSPSC:
46181504
NACRES:
NB.35
Productos recomendados
Materiales
Hypalon®
tamaño
8 (for 10 in. ports)
Características
reusable
envase
pack of 1 pair
fabricante / nombre comercial
Plas-Labs 800GH-8-10
Descripción general
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Información legal
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)