- Determining exhaust fumes exposure in chainsaw operations.
Determining exhaust fumes exposure in chainsaw operations.
Environmental pollution (Barking, Essex : 1987) (2016-09-12)
F Neri, C Foderi, A Laschi, F Fabiano, M Cambi, G Sciarra, M C Aprea, A Cenni, E Marchi
PMID27614911
ABSTRACT
The objective of this study was to investigate the inhalation exposure of forest operators to polycyclic aromatic hydrocarbons (PAHs) and BTEX (benzene, toluene, ethylbenzene and total xylenes) contained in the exhaust fumes released from chainsaws and to suggest possible countermeasures. The study was carried out in four silvicultural treatments (coppice clearcut, conifer thinning, conifer pruning, and sanitary cut), using three types of chainsaw fuel (normal two-stroke petrol mix and two alkylate fuels). Eighty personal air samples were collected; IOM samplers combined with Amberlite XAD-2 sorbent tubes were used for collecting PAHs and Radiello
Product Description
radiello™ BTEX/VOC Starter Kit, CS2 Desorption
radiello™ BTEX/VOC Starter Kit, Thermal Desorption
radiello™ Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello™ Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello™ Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello™ Filtration Kit, pk of 20
Supelco
radiello™ Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello™ Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello™ 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello™ On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello™ Clips, suspending Radiello triangular support plate, pk of 20
radiello™ Diffusive Bodies, white, configured for general use, pk of 20
radiello™ On-Field Thermometer, Reader, pk of 1
Supelco
radiello™ Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello™ Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello™ Hydrogen Sulfide Starter Kit
radiello™ Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello™ Barcode Labels, pk of 198
radiello™ Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello™ Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello™ Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello™ Anesthetic Gases and Vapor Sampler
radiello™ Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello™ Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello™ Outdoor Shelter, pk of 10
radiello™ Vertical Adapter, threaded for standard use, pk of 20
radiello™ Triangular Support Plate, pk of 20