Skip to Content
Merck
All Photos(1)

Key Documents

RAD168

radiello Cartridge Adsorbents

for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24
Pricing and availability is not currently available.

Agency

NIOSH 6015, 6016
OSHA ID-188

packaging

pk of 20

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

matrix

microporous PE impregnated with phosphoric acid

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling Ammonia (NH3)

Looking for similar products? Visit Product Comparison Guide

General description

Several different Radiello cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of Radiello thermal desorption (TD) cartridge adsorbents.

Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time.

Note: To conduct air sampling using the Radiello system, Radiello diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. Radiello Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Disclaimer

*Note: This product is not officially specified in the method, it may be suitable for use for the compound(s) listed in the method*

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Evaluation of airborne particles at the Alhambra monument in Granada, Spain
Horemans, B., et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 99 (2), 429-438 (2011)
Trends in atmospheric nitrogen and sulphur deposition in northern Belgium
Staelens, J., et al.
Atmospheric Environment, 49, 186-196 (2012)
Spatial and temporal variability of ammonia and other inorganic aerosol species
Day, D.E., et al.
Atmospheric Environment, 61, 490-498 (2012)

Articles

radiello® Diffusive Air Sampling Application - Ammonia

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Protocols

Learn more about radiello air sampler calibration solutions for aldehydes and H2s. We also offer many air sampler calibration kits.

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service