Skip to Content
Merck
All Photos(1)

Key Documents

RAD171

Supelco

radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide)

pkg of 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NB.24
Pricing and availability is not currently available.

packaging

pkg of 100 mL

application(s)

environmental

format

neat

General description

The H2S calibration standard contains a methylene blue concentrate that, once diluted 1:50 (v/v) with water, provides the same absorbance value of hydrogen sulfide at 665 nm at a concentration of 1.145 μg/mL sulfide ions. This concentration value is the highest absorbance value within the linear range of the spectrophotometer and can be used as a stock solution to prepare standards for the calibration curve. The concentrate is suitable for preparing up to 50 calibration curve and is stable for at least 1 year.

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Analyte

Description

    Methylene blue

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaqi Liu et al.
Cell & bioscience, 6, 33-33 (2016-05-26)
Excessive autophagy induced by extravagant oxidative stress is the main reason for diabetes-induced vascular endothelial cells dysfunction. Hydrogen sulfide (H2S) has anti-oxidative effects but its regulation on excessive autophagy of vascular endothelial cells is unclear. In this study, aorta of
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Hai-bin Dai et al.
BMC anesthesiology, 15, 119-119 (2015-08-19)
Both hydrogen sulphide (H2S) and mild hypothermia have been reported to prevent brain damage caused by reperfusion assault through regulating the N-methyl-D-aspartate receptor (NMDAR). However, the relationship between the two treatments and how they exert neuro-protective effects through NMDARs remain
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components

Articles

radiello® Diffusive Air Sampling Application - Hydrogen Sulfide

Related Content

Frequently Asked Questions - radiello® Products

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service