Key Documents
RAD195
radiello Clips
suspending Radiello triangular support plate, pk of 20
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
Pricing and availability is not currently available.
usage
suspending Radiello triangular support plate
packaging
pk of 20
manufacturer/tradename
Radiello
technique(s)
passive air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Rocco Rella et al.
The Science of the total environment, 487, 255-259 (2014-05-03)
The proofing treatment of the floor slabs with a solvent based material and successive treatment with epoxy resin resulted in significant air contamination in a primary school. The contamination caused illnesses in many occupants of school. The GC/MS investigations of
Anne Pennequin-Cardinal et al.
Talanta, 65(5), 1233-1240 (2008-10-31)
Radiello((R)) diffusive samplers filled with a thermally desorbable adsorbent (graphitised charcoal Carbograph 4) have been tested for the monitoring of BTEX. The sampling rates have been estimated under various controlled atmospheres in order to evaluate the effects of two factors
Armelle Vallat et al.
Phytochemistry, 66(13), 1540-1550 (2005-06-14)
Headspace volatiles from apple-bearing twigs were collected in the field with a Radiello sampler during three different diurnal periods over the complete fruit growing season. Analyses by thermal desorption-GC-MS identified a total of 62 compounds in changing quantities, including the
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Articles
Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocols
Learn more about radiello air sampler calibration solutions for aldehydes and H2s. We also offer many air sampler calibration kits.
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Related Content
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.