Merck
  • Fas Signaling Promotes Gastric Cancer Metastasis through STAT3-Dependent Upregulation of Fascin.
Fas Signaling Promotes Gastric Cancer Metastasis through STAT3-Dependent Upregulation of Fascin.
PloS one (2015-05-21)
Yunshan Yang, Qiyu Zhao, Zhijian Cai, Guoping Cheng, Ming Chen, Jiaoli Wang, Haijun Zhong
ABSTRACT
Fas signaling-activated signal transducers and activators of transcription 3 (STAT3) is required for Fascin upregulation. As an actin-bundling protein, Fascin can mediate gastric cancer (GC) cell migration. Gastric cancer AGS cells were treated with anti-Fas (5 μg/ml) for 2 h, in order to stimulate the activation of the Fas signaling. The in vitro migration of Fas signaling-activated AGS cells was assessed using Transwell chambers. The levels of Fascin and phosphorylated STAT3 were detected by Western blotting analyses. Nude mice were injected intravenously with AGS cells treated with anti-Fas or treated with STAT3 inhibitor without anti-Fas; tumor pulmonary metastases were measured. Fascin protein expression in tumor tissues was detected by immunohistochemistry. The Fas and Fascin mRNA levels in tumor tissues from patients with GC were measured by real-time PCR and their correlation was analyzed. The activation of Fas signaling promoted cell migration and resulted in STAT3-dependent Fascin upregulation in AGS cells. STAT3 enhanced Fascin levels in vivo. Fascin was the mediator of Fas signaling-induced AGS cell migration in vitro and in vivo. Furthermore, there was a positive correlation between Fas and Fascin mRNA levels in tumor tissues from GC patients. Fas signaling promotes GC metastasis through the STAT3/Fascin pathway, which may provide a new target for GC therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
S3I-201, ≥97% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fscn1
Sigma-Aldrich
MISSION® esiRNA, targeting human FSCN1
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O