Skip to Content
Merck
All Photos(7)

Key Documents

L4509

Sigma-Aldrich

Sodium dodecyl sulfate

Sodium dodecyl sulfate

ReagentPlus®, ≥98.5% (GC)

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$101.00
25 G
$145.00
100 G
$265.00
250 G
$499.00
500 G
$953.00
1 KG
$1,410.00

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$101.00
25 G
$145.00
100 G
$265.00
250 G
$499.00
500 G
$953.00
1 KG
$1,410.00

About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.21

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

description

anionic

product line

ReagentPlus®

Assay

≥98.5% (GC)

form

powder

mol wt

micellar avg mol wt 18,000

aggregation number

62

impurities

<2.0% water (Karl Fischer)

CMC

7-10 mM (20-25°C)

mp

204-207 °C (lit.)

transition temp

cloud point >100 °C

solubility

water: soluble

density

0.425 g/cm3

cation traces

Na: 7.0-9.0%

HLB

40

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dodecyl sulfate (SDS), or sodium lauryl sulfate, is widely recognized as an anionic surfactant within the organosulfate class of organic compounds. Its effectiveness as a detergent spans both ionic and anionic conditions. SDS plays a critical role as a protein denaturant, facilitating the solubilization and denaturation of proteins in procedures like protein extraction and SDS-PAGE. Its significance in the electrophoretic separation of proteins and lipids establishes its indispensability for the examination of these molecules in diverse biological experiments. In conclusion, Sodium dodecyl sulfate proves to be an indispensable element in the exploration of biological molecules within the realms of biochemical and cell biology research.

Application

Anionic detergent
Sodium dodecyl sulphate (SDS) has been used:
  • in the characterization of Enterobacteriaceae strain based on sensitivity to selective compounds[1]
  • to treat human skin in vitro and in vivo to evaluate the role of nuclear localization of 27-kDa heat shock protein (HSP27) in potential skin irritation[2]
  • in the decellularization of porcine heart valve tissue[3]
  • to treat sperm for in vitro induction of sperm decondensation[4]

Biochem/physiol Actions

Sodium dodecyl sulphate (SDS) helps to quickly disrupt the biological membranes. It is usually used as one of the major constituent of several reagents, that are used to purify nucleic acids. SDS can block the activity of RNase and deoxyribonuclease (DNase).[5]

Features and Benefits

  • Suitable for Cell Biology and Biochemical Research
  • High purity chemical suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid and reliable alternative to ISO 21528-1: 2004 for detection of Enterobacteriaceae
Joosten H, et al.
International Journal of Food Microbiology, 125(3), 344-346 (2008)
Resilient ribonucleases
RNA Methodologies: A Laboratory Guide for Isolation and Characterization, 32-45 (1993)
Sub-Micellar Concentration of Sodium Dodecyl Sulphate Prevents Thermal Denaturation Induced Aggregation of Plant Lectin, Jacalin
Lavanya V, et al
The Protein Journal, 36(1), 17-27 (2017)
Utility of light scatter in the morphological analysis of sperm
Zucker RM, et al.
Cytometry, 39-47 (1992)
Minimally immunogenic decellularized porcine valve provides in situ recellularization as a stentless bioprosthetic valve.
Iwai S, et al.
Journal of Artificial Organs : The Official Journal of the Japanese Society for Artificial Organs, 10(1), 29-35 (2007)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service