Skip to Content
Merck
All Photos(2)

Key Documents

P4864

Sigma-Aldrich

Propidium iodide solution

≥94% purity, solution (1.0 mg/ml in water)

Synonym(s):

3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide, 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium, diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34I2N4
CAS Number:
Molecular Weight:
668.39
Beilstein:
3843838
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Propidium iodide solution,

Assay

≥94%

Quality Level

form

liquid

technique(s)

titration: suitable

color

faint orange to very dark orange, and Faint Red to Very Dark Red and Faint Purple to Very Dark Purple and Orange-Red and Red-Orange

solubility

water: 1 mg/mL

εmax

5400-6400 at 491-495 nm in phosphate buffer

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[I-].[I-].CC[N+](C)(CC)CCC[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C27H33N4.2HI/c1-4-31(3,5-2)17-9-16-30-26-19-22(29)13-15-24(26)23-14-12-21(28)18-25(23)27(30)20-10-7-6-8-11-20;;/h6-8,10-15,18-19,29H,4-5,9,16-17,28H2,1-3H3;2*1H/q+1;;/p-1

InChI key

XJMOSONTPMZWPB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Propidium iodide solution is used to assess neuronal death.

Application

Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Suitable for use in fluorescence microscopy, confocal laser scanning microscopy, flow cytometry, and fluorometry. Used as a nuclear counterstain.
Propidium iodide solution has been used:
  • in staining DNA
  • in the viability staining method to study islet viability
  • to visualize F-actins and nuclei in mesenchymal stem cell sheets

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paternal cyclophosphamide exposure induces the formation of functional micronuclei during the first zygotic division
Grenier L, et al.
PLoS ONE, 6(11), e27600-e27600 (2011)
Direct intramyocardial injection of mesenchymal stem cell sheet fragments improves cardiac functions after infarction
Wang CC, et al.
Cardiovascular Research, 77(3), 515-524 (2007)
Chung-Chi Wang et al.
Cardiovascular research, 77(3), 515-524 (2007-11-17)
Cell transplantation is a promising approach for patients with myocardial infarction. However, following injection, retention of the transplanted cells in the injected area remains a central issue, which can be deleterious to cell transplantation therapy. We hypothesized that the use
Stroke E-Book: Pathophysiology Diagnosis and Management, 79-79 (2011)
Variation in human islet viability based on different membrane integrity stains
Barnett MJ, et al.
Cell Transplantation, 13(5), 481-488 (2004)

Articles

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Related Content

Nancy-520 for DNA Detection and Quantitation

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service