Skip to Content
Merck
All Photos(4)

Key Documents

L6026

Sigma-Aldrich

Sodium dodecyl sulfate

BioXtra, ≥99.0% (GC)

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$89.30
250 G
$308.00
1 KG
$924.00

$89.30

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$89.30
250 G
$308.00
1 KG
$924.00

About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.21

$89.30

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 01, 2025Details


Request a Bulk Order

biological source

synthetic (oragnic)

Quality Level

description

anionic

product line

BioXtra

Assay

≥99.0% (GC)

form

powder

mol wt

micellar avg mol wt 18,000

aggregation number

62

impurities

Insoluble matter, passes filter test

loss

≤0.5% loss on drying, HV, 20-25 °C

CMC

7-10 mM (20-25°C)

mp

204-207 °C (lit.)

transition temp

cloud point >100 °C

solubility

H2O: 0.1 M, clear, colorless

density

0.496 g/cm3

anion traces

chloride (Cl-): ≤0.05%
phosphate (PO43-): ≤0.0001%

cation traces

Al: ≤0.0005%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.02%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.05 at 280 in H2O at 0.1 M
≤0.08 at 260 in H2O at 0.1 M

HLB

40

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dodecyl sulfate (SDS) is a well-known anionic detergent widely used in scientific research to denature proteins′ secondary and tertiary structures. It consists of a 12-carbon non-polar chain with a negatively charged sulfate group at the end. SDS is instrumental in denaturing and solubilizing proteins, making it a fundamental component of procedures like polyacrylamide gel electrophoresis, where it helps determine protein molecular weights.Moreover, SDS acts as an organic sodium salt and serves dual roles as a detergent and a protein denaturant. Its distinctive feature is its anionic surfactant nature, characterized by a negative charge. This charge fosters interactions with proteins and lipids, leading to the creation of micellar structures. These micelles are essential for SDS′s capacity to effectively solubilize and stabilize proteins and lipids, enabling their detailed study and manipulation in scientific research. SDS′s amphipathic properties, stemming from a long-chain aliphatic group with an attached negatively charged sulfate, make it a valuable detergent for various biomolecular experiments and studies.

Application

Anionic detergent
Sodium dodecyl sulfate has been used as a:
  • surfactant to evaluate cytochrome c affinity by using surface plasmon resonance[1]
  • surfactant in particle enhanced immunoassays to inhibit aggregation of particles and to enhance single microsphere coupling[2]
  • biofilm-disrupting agent[3]

Features and Benefits

  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Tested for trace levels of Anions and Cations
  • High-purity product ideal for biochemical research purposes

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomedical Devices and Their Applications (2013)
Evaluation of cytochrome c affinity to anionic phospholipids by means of surface plasmon resonance
Stepanov G, et al.
Febs Letters, 583(1), 97-100 (2009)
Lonnie J Lucas et al.
Colloids and surfaces. B, Biointerfaces, 49(2), 106-111 (2006-04-20)
Manufacturers of latex immunoassays have typically added surfactants to improve detection sensitivity and prevent non-specific aggregation of microspheres, which may cause both false positives and negatives during diagnostic testing. There is also growing interest in conducting immunoassays in smaller volumes
Brandon M Childers et al.
Infection and immunity, 81(11), 4112-4120 (2013-08-28)
Biofilm formation and persistence are essential components for the continued survival of pathogens inside the host and constitute a major contributor to the development of chronic wounds with resistance to antimicrobial compounds. Understanding these processes is crucial for control of
Shane Stegeman et al.
Endocrine-related cancer, 22(2), 265-276 (2015-02-12)
The oncogene MDM4, also known as MDMX or HDMX, contributes to cancer susceptibility and progression through its capacity to negatively regulate a range of genes with tumour-suppressive functions. As part of a recent genome-wide association study it was determined that

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service