Merck
  • Targeting of FLT3-ITD kinase contributes to high selectivity of imidazoacridinone C-1311 against FLT3-activated leukemia cells.
Targeting of FLT3-ITD kinase contributes to high selectivity of imidazoacridinone C-1311 against FLT3-activated leukemia cells.
Biochemical pharmacology (2015-04-22)
Anna Skwarska, Ewa Augustin, Michał Beffinger, Anita Wojtczyk, Sonia Konicz, Katarzyna Laskowska, Joanna Polewska
ABSTRACT
Drugs targeting receptor tyrosine kinase FLT3 are of particular interest since activating FLT3-internal tandem duplication (ITD) mutations abundantly occur in fatal acute myeloid leukemias (AMLs). Imidazoacridinone C-1311, a DNA-reactive inhibitor of topoisomerase II, has been previously shown to be a potent and selective inhibitor of recombinant FLT3. Here, we expand those findings by studying its effect on leukemia cells with wild-type FLT3, FLT3-ITD mutant and no FLT3 receptor. While brief C-1311 exposure blocked wild-type and FLT3-ITD activity, profound and sustained inhibition was achieved only for FLT3-ITD mutants. C-1311 inhibited FLT3 downstream pathways (MAPK and AKT) independent of FLT3 status, yet translation to decreased viability was significant in FLT3-ITD cells. RNA interference against FLT3-ITD reduced cytotoxic effect and apoptosis induced by C-1311, indicating selective inhibition of FLT3-ITD crucial for high efficacy of drug against activated leukemia cells. Cellular responses in treated FLT3-ITD mutants included G1 and G2/M phase arrest, moderate inhibition of Bcl-2, caspase-3 activation, PARP cleavage, and depolarization of mitochondria. Consistent with selective decrease in FLT3-ITD activity, C-1311 remarkably reduced antiapoptotic survivin mRNA and protein expression, correlating well with enhanced apoptosis of FLT3-ITD cells. No survivin decrease and respectively lower level of apoptosis was found in wild-type and null-FLT3 cells. Combination of C-1311 with cytarabine or doxorubicin again showed distinct synergistic activity in FLT3-ITD-positive cells. The ability of C-1311 to selectively target constitutively active FLT3, suggests a favorable therapeutic index for AML carrying FLT3-ITD mutations. Thus further preclinical and clinical studies addressing its potency against FLT3-ITD kinase is well justified.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Flt3
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)