Skip to Content
Merck
All Photos(3)

Key Documents

450243

Sigma-Aldrich

Sodium orthovanadate

99.98% trace metals basis

Synonym(s):

Trisodium vanadate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$97.30
50 G
$260.00

$97.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$97.30
50 G
$260.00

About This Item

Linear Formula:
Na3VO4
CAS Number:
Molecular Weight:
183.91
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$97.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.98% trace metals basis

form

solid

reaction suitability

core: vanadium
reagent type: catalyst

mp

850-866 °C (lit.)

SMILES string

[Na+].[Na+].[Na+].[O-][V]([O-])([O-])=O

InChI

1S/3Na.4O.V/q3*+1;;3*-1;

InChI key

IHIXIJGXTJIKRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium orthovanadate (Na3VO4) can be used as a metal precursor in the synthesis of:
  • Vanadium-MCM-48 (V-MCM-48) by anion-exchange method. V-MCM-48 is used as a catalyst in oxidation of styrene to benzaldehyde.[1]
  • Nanorods of vanadium oxide particles by glycothermal method.[2]
  • Yttrium vanadate (YVO4) doped with colloidal europium.[3]

Biochem/physiol Actions

An inhibitor of protein tyrosine phosphatases, alkaline phosphatases and ATPases.
Inhibits ATPase, alkaline phosphatase and tyrosine phosphatase. Decavanadate, which is formed at acidic pH, inhibits inositol 1,4,5-trisphosphate (IP3)-induced Ca2+ release from endocrine cells and blocks IP3 binding to its receptor in brain tissue.

Packaging

Exhibits various biological and physiological effects in the human body[4][5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of TM-MCM-48 (TM= Mn, V, Cr) and their catalytic activity in the oxidation of styrene.
Gomez S, et al.
J. Catal., 233(1), 60-67 (2005)
Synthesis and characterizations of YVO4: Eu colloids.
Huignard A, et al.
Chemistry of Materials, 14(5), 2264-2269 (2002)
Glycothermal synthesis of assembled vanadium oxide nanostructures for gas sensing.
Fu H, et al.
Journal of Nanoparticle Research, 14(6), 871-871 (2012)
Yu Hasegawa et al.
The Journal of pharmacology and experimental therapeutics, 317(2), 875-881 (2006-02-08)
Vanadium is widely distributed in the environment and exhibits various biological and physiological effects in the human body. We previously documented the neuroprotective effect of sodium orthovanadate (SOV) against in rodents i.v. injected with 2 ml/kg 50 mM SOV just
Magdalena Malm et al.
Biotechnology journal, 9(9), 1215-1222 (2014-03-29)
Emerging strategies in cancer biotherapy include the generation and application of bispecific antibodies, targeting two tumor-associated antigens for improved tumor selectivity and potency. Here, an alternative format for bispecific molecules was designed and investigated, in which two Affibody molecules were

Questions

1–6 of 6 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the solubility of Product 450243, sodium orthovandate, and how do I store the solution?

    1 answer
    1. Solutions prepared as 1 mM in water (or higher if desired) and adjusted to pH 10 can be aliquotted in plastic tubes and stored at -20°C. To ensure the presence of monomers, solutions should be heated to boiling until translucent and pH re-adjusted to 10.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. What is Product 450243, Sodium orthovanadate, typically used for?

    1 answer
    1. This product can be used as a catalyst in chemical synthesis. It also utilized as an inhibitor of ATPase, alkaline phosphatase and tyrosine phosphatase.

      Helpful?

  6. For Product 450243, Sodium orthovanadate, what does metal basis purity mean?

    1 answer
    1. This product's purity is determined by ICP analysis. Trace metal impurities are analyzed, quantified and reported on the certificate of analysis for each lot/batch of product. The total metallic impurities are subtracted from 100% to result in the purity percentage based on metals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service