Merck
  • Ascochlorin, an isoprenoid antibiotic inhibits growth and invasion of hepatocellular carcinoma by targeting STAT3 signaling cascade through the induction of PIAS3.
Ascochlorin, an isoprenoid antibiotic inhibits growth and invasion of hepatocellular carcinoma by targeting STAT3 signaling cascade through the induction of PIAS3.
Molecular oncology (2015-01-28)
Xiaoyun Dai, Kwang Seok Ahn, Chulwon Kim, Kodappully Sivaraman Siveen, Tina H Ong, Muthu K Shanmugam, Feng Li, Jizhong Shi, Alan Prem Kumar, Ling Zhi Wang, Boon Cher Goh, Junji Magae, Kam M Hui, Gautam Sethi
ABSTRACT
Deregulated activation of oncogenic transcription factors such as signal transducer and activator of transcription 3 (STAT3) plays a pivotal role in proliferation and survival of hepatocellular carcinoma (HCC). Thus, agents which can inhibit STAT3 activation may have an enormous potential for treatment of HCC patients. Hence, in the present report, we investigated the effect of ascochlorin (ASC), an isoprenoid antibiotic on STAT3 activation cascade in various HCC cell lines and orthotopic mouse model. We observed that ASC could substantially inhibit both constitutive and IL-6/EGF inducible STAT3 activation as well as reduce its DNA binding ability. ASC increased the expression of protein inhibitor of activated STAT3 (PIAS3) which could bind to STAT3 DNA binding domain and thereby down-regulate STAT3 activation. Deletion of PIAS3 gene by siRNA abolished the ability of ASC to inhibit STAT3 activation and induce apoptosis in HCC cells. ASC also modulated the expression of diverse STAT3-regulated oncogenic gene products. Finally, when administered intraperitoneally, ASC also inhibited tumor growth in an orthotopic HCC mouse model and reduced STAT3 activation in tumor tissues. Overall our results indicate that ASC mediates its anti-tumor effects predominantly through the suppression of STAT3 signaling cascade, and can form the basis of novel therapy for HCC patients.
Sigma-Aldrich
N-Iodosuccinimide, 95%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
MISSION® esiRNA, targeting human PIAS3
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)