Merck
MicroRNA let-7b regulates genomic balance by targeting Aurora B kinase.
Molecular oncology (2015-02-17)
Jenni Heidi Eveliina Mäki-Jouppila, Sofia Pruikkonen, Mahesh Balasaheb Tambe, Miriam Ragle Aure, Tuuli Halonen, Anna-Leena Salmela, Leena Laine, Anne-Lise Børresen-Dale, Marko Johannes Kallio
ABSTRACT
The let-7 microRNA (miRNA) family has been implicated in the regulation of diverse cellular processes and disease pathogenesis. In cancer, loss-of-function of let-7 miRNAs has been linked to tumorigenesis via increased expression of target oncogenes. Excessive proliferation rate of tumor cells is often associated with deregulation of mitotic proteins. Here, we show that let-7b contributes to the maintenance of genomic balance via targeting Aurora B kinase, a key regulator of the spindle assembly checkpoint (SAC). Our results indicate that let-7b binds to Aurora B kinase 3'UTR reducing mRNA and protein expression of the kinase. In cells, excess let-7b induced mitotic defects characteristic to Aurora B perturbation including increased rate of polyploidy and multipolarity, and premature SAC inactivation that leads to forced exit from chemically induced mitotic arrest. Moreover, the frequency of aneuploid HCT-116 cells was significantly increased upon let-7b overexpression compared to controls. Interestingly, together with a chemical Aurora B inhibitor, let-7b had an additive effect on polyploidy induction in HeLa cells. In breast cancer patients, reduced let-7b expression was found to be associated with increased Aurora B expression in grade 3 tumors. Furthermore, let-7b was found downregulated in the most aggressive forms of breast cancer determined by clinicopathological parameters. Together, our findings suggest that let-7b contributes to the fidelity of cell division via regulation of Aurora B. Moreover, the loss of let-7b in aggressive tumors may drive tumorigenesis by up-regulation of Aurora B and other targets of the miRNA, which further supports the role of let-7b in tumor suppression.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)