Skip to Content
Merck
All Photos(4)

Key Documents

553468

Sigma-Aldrich

Tris(tert-butoxy)silanol

99.999%

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3CO)3SiOH
CAS Number:
Molecular Weight:
264.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999%

form

solid

bp

205-210 °C (lit.)

mp

63-65 °C (lit.)

SMILES string

CC(C)(C)O[Si](O)(OC(C)(C)C)OC(C)(C)C

InChI

1S/C12H28O4Si/c1-10(2,3)14-17(13,15-11(4,5)6)16-12(7,8)9/h13H,1-9H3

InChI key

HLDBBQREZCVBMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(tert-butoxy)silanol can react with various metal alkyl amides to act as precursors for vapor deposition metal silicates. It also acts as a suitable precursor for deposition of silica.

Application

Tris(tert-alkoxy)silanols reacts with tetrakis(dimethylamino)-hafnium vapor(Hf(N(CH3)2)4) for vapor phase deposition of hafnium silicate glass films. Tris(tert-butoxy)silanol is used for atomic layer deposition (ALD) of highly conformal layers of amorphous silicon dioxide and aluminum oxide nanolaminates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Rapid vapor deposition of highly conformal silica nanolaminates.
Hausmann D, et al.
Science, 298(5592), 402-406 null
Rapid SiO2 Atomic Layer Deposition Using Tris(tert-pentoxy)silanol.
Burton B, et al.
Chemistry of Materials, 20, 7031-7043 (2008)
Vapor deposition of metal oxides and silicates: Possible gate insulators for future microelectronics.
Gordon G, et al.
Chemistry of Materials, 13(8), 2463-2464 (2001)
Jonathan Stewart et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(3), 428-436 (2014-09-27)
The oncogenic role of WNT is well characterized. Wntless (WLS) (also known as GPR177, or Evi), a key modulator of WNT protein secretion, was recently found to be highly overexpressed in malignant astrocytomas. We hypothesized that this molecule may be
Pedro A Dionísio et al.
Neurobiology of aging, 36(1), 228-240 (2014-12-03)
Alzheimer's disease (AD) is a neurodegenerative disorder hallmarked by the accumulation of extracellular amyloid-β (Aβ) peptide and intraneuronal hyperphosphorylated tau, as well as chronic neuroinflammation. Tauroursodeoxycholic acid (TUDCA) is an endogenous anti-apoptotic bile acid with potent neuroprotective properties in several

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service