Merck
  • Development of drug-in-adhesive patch formulations for transdermal delivery of fluoxetine: In vitro and in vivo evaluations.
Development of drug-in-adhesive patch formulations for transdermal delivery of fluoxetine: In vitro and in vivo evaluations.
International journal of pharmaceutics (2015-04-12)
Eunjae Jung, Eun Young Lee, Hoo-Kyun Choi, Sang-Jun Ban, Seung-Hyuk Choi, Jung Sun Kim, In-Soo Yoon, Dae-DuK Kim
ABSTRACT
The aim of this study was to develop drug-in adhesive (DIA) patch formulations for the transdermal delivery of fluoxetine (FX). The DIA patch formulations containing FX were prepared and optimized with various pressure sensitive adhesives, drug contents and enhancers. Among the various formulations, DuroTak 87-502B-based patch formulation containing 20% (w/w) FX with no enhancer was selected for in vivo pharmacokinetic study based on in vitro permeation studies using hairless mouse, rat and human cadaver skins. The Cmax of FX after the transdermal administration of the optimized patch formulation in rats was 52.38 ng/ml, and plasma concentration of FX was maintained for 36 h. Moreover, the predicted human Css (55.79 ng/ml) and Cmax (27.35 ng/ml) were in good agreement with the reported plasma levels (15-55 ng/ml) of oral FX in clinical applications. These results suggest that the optimized patch formulation could be further developed for clinical applications, providing the therapeutic plasma level of FX over an extended period. To the best of our knowledge, our results are the first reported in vitro and in vivo data on the preparation and optimization of FX-loaded DIA patch, showing its feasibility as an effective transdermal delivery system for FX.
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Di(ethylene glycol) ethyl ether, ≥99%
Sigma-Aldrich
Oleic acid, ≥99% (GC)