Skip to Content
Merck
All Photos(1)

Key Documents

172472

Sigma-Aldrich

Isopropyl myristate

Isopropyl myristate

98%

Synonym(s):

Isopropyl tetradecanoate, Myristic acid isopropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$55.50
1 L
$260.00

$55.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$55.50
1 L
$260.00

About This Item

Linear Formula:
CH3(CH2)12COOCH(CH3)2
CAS Number:
Molecular Weight:
270.45
Beilstein:
1781127
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39022421
PubChem Substance ID:
NACRES:
NA.22

$55.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

193 °C/20 mmHg (lit.)

mp

~3 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C17H34O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-17(18)19-16(2)3/h16H,4-15H2,1-3H3

InChI key

AXISYYRBXTVTFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

IPM is a fatty acid ester that can be produced by the esterification of isopropyl alcohol and myristic acid.[1]

Application

Isopropyl myristate was used as oil phase in the preparation of formulations[2]. It was also used as oil/water-type tacrolimus (FK506) solvent[3].

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Neslihan Ustündağ Okur et al.
Chemical & pharmaceutical bulletin, 62(2), 135-143 (2014-02-05)
Naproxen (Np) is an example of a non-steroidal anti-inflammatory drug (NSAID) commonly used for the reduction of pain and inflammation. In order to develop an alternative formulation for the topical administration of Np, microemulsions were evaluated as delivery vehicles. Four
Green synthesis of isopropyl myristate in novel single phase medium Part I: Batch optimization studies
Vadgama R, et al.
Biotechnology reports (Amsterdam, Netherlands), 8, 133-137 (2015)
Masayuki Yamanaka et al.
International journal of pharmaceutics, 464(1-2), 19-26 (2014-01-25)
We developed an oil/water-type tacrolimus (FK506) cream formulation as an alternative to Protopic ointment for atopic dermatitis treatment. We determined the effects of solvents used in topical preparations on FK506 solubility and stability, and evaluated FK506 transdermal absorption into rat
Timothy S McClintock et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(47), 15669-15678 (2014-11-21)
Our understanding of mammalian olfactory coding has been impeded by the paucity of information about the odorant receptors (ORs) that respond to a given odorant ligand in awake, freely behaving animals. Identifying the ORs that respond in vivo to a
Santiago Delgado-Tirado et al.
Scientific reports, 10(1), 20554-20554 (2020-12-02)
Proliferative vitreoretinopathy (PVR) is the leading cause of retinal detachment surgery failure. Despite significant advances in vitreoretinal surgery, it still remains without an effective prophylactic or therapeutic medical treatment. After ocular injury or retinal detachment, misplaced retinal cells undergo epithelial

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. I need to know the expiry date of batch STBK7141 of product 172472 please

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Customers should routinely inspect products with no expiration date or recommended retest date to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

Active Filters

  1. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service