Merck
  • Effects of artemisinin antimalarials on Cytochrome P450 enzymes in vitro using recombinant enzymes and human liver microsomes: potential implications for combination therapies.
Effects of artemisinin antimalarials on Cytochrome P450 enzymes in vitro using recombinant enzymes and human liver microsomes: potential implications for combination therapies.
Xenobiotica; the fate of foreign compounds in biological systems (2014-01-10)
Therese Ericsson, Jesper Sundell, Angelica Torkelsson, Kurt-Jürgen Hoffmann, Michael Ashton
ABSTRACT
1. Cytochrome P450 enzyme system is the most important contributor to oxidative metabolism of drugs. Modification, and more specifically inhibition, of this system is an important determinant of several drug-drug interactions (DDIs). 2. Effects of the antimalarial agent artemisinin and its structural analogues, artemether, artesunate and dihydroartemisinin, on seven of the major human liver CYP isoforms (CYP1A2, 2A6, 2B6, 2C9, 2C19, 2D6 and 3A4) were evaluated using recombinant enzymes (fluorometric assay) and human liver microsomes (LC-MS/MS analysis). Inhibitory potency (IC50) and mechanisms of inhibition were evaluated using nonlinear regression analysis. In vitro-in vivo extrapolation using the [I]/Ki ratio was applied to predict the risk of DDI in vivo. 3. All compounds tested inhibited the enzymatic activity of CYPs, mostly through a mixed type of inhibition, with CYP1A2, 2B6, 2C19 and 3A4 being affected. A high risk of interaction in vivo was predicted if artemisinin is coadministrated with CYP1A2 or 2C19 substrates. 4. With respect to CYP1A2 inhibition in vivo by artemisinin compounds, our findings are in line with previously published data. However, reported risks of interaction may be overpredicted and should be interpreted with caution.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride, 97%
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Supelco
Umbelliferone, analytical standard
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, Molecular Biology, ≥99.0% (KT)
Sigma-Aldrich
7-Benzyloxy-4-trifluoromethylcoumarin, ≥99% (HPLC), solid
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Chlorpropamide, analytical standard, ≥97%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
7-Ethoxy-4-(trifluoromethyl)coumarin, ≥98% (TLC)
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder