Skip to Content
Merck
All Photos(1)

Key Documents

T0891

Supelco

Tolbutamide

analytical standard

Synonym(s):

1-Butyl-3-(4-methylphenylsulfonyl)urea, N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$52.40
100 G
$135.00

$52.40


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$52.40
100 G
$135.00

About This Item

Empirical Formula (Hill Notation):
C12H18N2O3S
CAS Number:
Molecular Weight:
270.35
Beilstein:
1984428
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$52.40


Estimated to ship onMay 24, 2025


Request a Bulk Order

biological source

synthetic (organic)

Quality Level

grade

analytical standard

Assay

97.0-103.0%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

ethanol: 50 mg/mL, clear, colorless

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCCNC(=O)NS(=O)(=O)c1ccc(C)cc1

InChI

1S/C12H18N2O3S/c1-3-4-9-13-12(15)14-18(16,17)11-7-5-10(2)6-8-11/h5-8H,3-4,9H2,1-2H3,(H2,13,14,15)

InChI key

JLRGJRBPOGGCBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anti-diabetic agent. Metabolized by CYP2C9 (tolbutamide hydroxylase).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin S C Hamming et al.
Diabetes, 59(7), 1686-1693 (2010-04-24)
The sodium-calcium exchanger isoform 1 (NCX1) regulates cytoplasmic calcium (Ca(2+)(c)) required for insulin secretion in beta-cells. NCX1 is alternatively spliced, resulting in the expression of splice variants in different tissues such as NCX1.3 and -1.7 in beta-cells. As pharmacological inhibitors
Ruurdtje Hoekstra et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(3), 562-567 (2012-12-15)
The human liver cell line HepaRG has been recognized as a promising source for in vitro testing of metabolism and toxicity of compounds. However, currently the hepatic differentiation of these cells relies on exposure to dimethylsulfoxide (DMSO), which, as a
Ioannis Eleftherianos et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(29), 12024-12029 (2011-07-02)
The effects of the cellular environment on innate immunity remain poorly characterized. Here, we show that in Drosophila ATP-sensitive potassium channels (K(ATP)) mediate resistance to a cardiotropic RNA virus, Flock House virus (FHV). FHV viral load in the heart rapidly
Jean-Claude Henquin et al.
Diabetes, 62(5), 1689-1696 (2013-01-01)
Congenital hyperinsulinism causes persistent hypoglycemia in neonates and infants. Most often, uncontrolled insulin secretion (IS) results from a lack of functional K(ATP) channels in all β-cells or only in β-cells within a resectable focal lesion. In more rare cases, without
Jingwen Gao et al.
Journal of ethnopharmacology, 145(2), 566-572 (2012-12-05)
Cooked rhubarb and wine processed rhubarb are the processed rhubarbs of raw rhizomes from Rheum palmatum L., Rheum tanguticum Maxim. ex Balf. or Rheum officinale Baill. They are clinically used in traditional Chinese medicine to compose anti-diabetic formulas and remove

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service