Skip to Content
Merck
All Photos(3)

Documents

P8281

Sigma-Aldrich

Potassium phosphate dibasic

reagent grade, ≥98.0%

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070212
PubChem Substance ID:
NACRES:
NA.25

grade

reagent grade

Quality Level

Assay

≥98.0%

form

powder

pH

8.7-9.3 (25 °C, 50 g/L)

solubility

water: 200 mg/mL, clear to slightly hazy, colorless

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Nanoparticles and Chemical Inducers: A Sustainable Shield against Onion White Rot.: This research discusses the effectiveness of potassium phosphate dibasic as a chemical inducer in developing sustainable agricultural practices to combat fungal infections in crops. The study provides insights into the potential of integrating chemical inducers with nanoparticles to enhance crop resilience and yield (Elenany AM et al., 2024).

  • Development of Simultaneous Analytical Method for Imidazolinone Herbicides from Livestock Products by UHPLC-MSMS.: This study explores the use of potassium phosphate dibasic in chromatography media to enhance the detection of herbicide residues in food products, ensuring food safety and regulatory compliance. The research underscores the critical role of advanced analytical methods in maintaining the integrity of the food supply (Heo HM et al., 2022).

  • High-Throughput Method for Natural Organic Matter Hydrophobicity Assessment Using an Ionic Liquid-Based Aqueous Two-Phase System.: Employing potassium phosphate dibasic, this study enhances the understanding of natural organic matter′s behavior in aquatic environments, crucial for water treatment and environmental remediation strategies. The method offers a novel approach to evaluating water quality and treatment efficiencies (Wang J et al., 2021).

  • Using aquatic vegetation to remediate nitrate, ammonium, and soluble reactive phosphorus in simulated runoff.: This article examines the role of potassium phosphate dibasic in environmental science, particularly its use in studies simulating the nutrient remediation potential of aquatic plants. The findings contribute to the development of natural treatment systems for nutrient-rich waters (Moore MT et al., 2016).

  • β-lactoglobulin stabilized nanemulsions--Formulation and process factors affecting droplet size and nanoemulsion stability.: Highlighting the application of potassium phosphate dibasic in pharmaceutical formulations, this study delves into the stabilization mechanisms of protein-based nanemulsions, crucial for the delivery of therapeutic agents. The research aids in optimizing formulations for enhanced drug delivery systems (Ali A et al., 2016).


Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayse Karatug Kacar et al.
The Journal of pharmacy and pharmacology, 72(4), 592-606 (2020-01-25)
The purpose of this study was to prepare targeted cancer therapy formulation against insulinoma INS-1 cells and to study its effect on cell death with related mechanisms in vitro. Polylactide-co-glycolide (PLGA) nano-micelles were used for preparation of esculetin nano-formulation (nano-esculetin). The
Ming Ye et al.
Biosensors & bioelectronics, 73, 208-213 (2015-06-17)
Several volatile organic compounds (VOCs) are released from human breath or skin. Like chemical substances in blood or urine, some of these vapors can provide valuable information regarding the state of the human body. A highly sensitive acetone biochemical gas
Pamela Gamba et al.
PLoS genetics, 11(6), e1005047-e1005047 (2015-06-26)
Gene expression can be highly heterogeneous in isogenic cell populations. An extreme type of heterogeneity is the so-called bistable or bimodal expression, whereby a cell can differentiate into two alternative expression states. Stochastic fluctuations of protein levels, also referred to
Kenshi Hayakawa et al.
Journal of bioscience and bioengineering, 120(5), 532-538 (2015-04-29)
S-Adenosyl-L-methionine (SAM) is a major biological methyl group donor, and is used as a nutritional supplement and prescription drug. Yeast is used for the industrial production of SAM owing to its high intracellular SAM concentrations. To determine the regulation mechanisms
Audrey Beaussart et al.
ACS nano, 8(10), 10723-10733 (2014-10-07)
A variety of bacterial pathogens use nanoscale protein fibers called type IV pili to mediate cell adhesion, a primary step leading to infection. Currently, how these nanofibers respond to mechanical stimuli and how this response is used to control adhesion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service