Skip to Content
Merck
All Photos(1)

Key Documents

B5057

Sigma-Aldrich

7-Benzyloxy-4-trifluoromethylcoumarin

≥99% (HPLC), solid

Synonym(s):

7-BFC

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$882.00

$882.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
5 MG
$882.00

About This Item

Empirical Formula (Hill Notation):
C17H11F3O3
Molecular Weight:
320.26
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.47

$882.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Product Name

7-Benzyloxy-4-trifluoromethylcoumarin, ≥99% (HPLC), solid

Quality Level

Assay

≥99% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

FC(F)(F)C1=CC(=O)Oc2cc(OCc3ccccc3)ccc12

InChI

1S/C17H11F3O3/c18-17(19,20)14-9-16(21)23-15-8-12(6-7-13(14)15)22-10-11-4-2-1-3-5-11/h1-9H,10H2

InChI key

WVKLERKKJXUPIK-UHFFFAOYSA-N

Gene Information

Application

7-Benzyloxy-4-trifluoromethylcoumarin has been used:

  • in the SBAD2-3x-AAGLY hepatocyte-like cell (HLC) functional assay to measure the cytochrome P450 3A4 (CYP3A4)-dependent activity
  • in O-dealkylation reaction to determine the activity of CYP3A
  • to measure the activity of CYP3A-mediated 7-benzyloxy-4-trifluoromethylcoumarin O-debenzylase activity (BFCOD)

Biochem/physiol Actions

Metabolized primarily by cytochrome P450 isozymes CYP1A2 and CYP3A4.

Legal Information

Supplied by Gentest Corp., U.S. Patent Nos. 6,207,404.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiannan Ding et al.
Environmental pollution (Barking, Essex : 1987), 238, 1-9 (2018-03-13)
While the presence of microplastics (MPs) in marine environments has been detected worldwide, the importance of MPs pollution in freshwater environments has also been emphasized in recent years. However, the body of knowledge regarding the biological effects of MPs on
Jiannan Ding et al.
Chemosphere, 148, 21-31 (2016-01-23)
The aim of this study was to evaluate the interactive effects of fluoxetine (FLU), roxithromycin (ROX) and propranolol (PRP) on the bioaccumulation and biochemical responses in the crucian carp Carassius auratus. After 7 days of binary exposure (ROX + FLU
Manami Nishiwaki et al.
Scientific reports, 10(1), 17503-17503 (2020-10-17)
Hepatocytes are an important tool for in vitro toxicology testing. In addition to primary cultures, a limited number of immortalized cell lines have been developed. We here describe a new cell line, designated as HepaMN, which has been established from
Vamshi K Manda et al.
Phytotherapy research : PTR, 31(12), 1935-1945 (2017-10-27)
Kratom (Mitragyna speciosa), a native herb of Southeast Asia, is widely known for its psychoactive properties. Recent increase in the use of kratom as a recreational drug has increased the risk of its interaction with conventional drugs if taken concomitantly.
Risto O Juvonen et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(9), 1015-1024 (2018-10-03)
Cytochrome P450 (CYP) enzymes constitute an essential xenobiotic metabolizing system that regulates the elimination of lipophilic compounds from the body. Convenient and affordable assays for CYP enzymes are important for assessing these metabolic pathways. In this study, 10 novel profluorescent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service