Merck
  • Sensitive naked eye detection of hydrogen sulfide and nitric oxide by aza-BODIPY dyes in aqueous medium.
Sensitive naked eye detection of hydrogen sulfide and nitric oxide by aza-BODIPY dyes in aqueous medium.
Analytical chemistry (2014-08-26)
Nagappanpillai Adarsh, Megha S Krishnan, Danaboyina Ramaiah
ABSTRACT
With an objective to develop optical probes for biologically important anions and neutral molecules, we synthesized three novel NIR absorbing aza-BODIPY derivatives, 3a-3c, and have systematically tuned their photophysical properties by changing the peripheral substitution. A profound red-shift was observed in the absorption and fluorescence spectra of the aza-BODIPY dyes with the change in substitution from azido (3a) to amino (3b) to dimethylamino (3c) groups. Theoretical calculations of 3a-3c showed a consistent decrease in bandgap, which supports the observed spectral changes. The study of their interactions with various analytes revealed that the azido-aza-BODIPY 3a selectively interacts with hydrogen sulfide (H2S) when compared to other molecules. Uniquely, the detection of H2S can be visualized through a change in color from bright blue to purple with a detection limit of 0.5 ppm. The sensitivity of the probe was observed to be ~20-fold higher than the allowed exposure limits of H2S as defined by EPA (10 ppm). The aza-BODIPY derivative 3b, on the other hand, exhibited selective interactions with nitrite ions (NO2(-)) and nitric oxide (NO) in aqueous medium through a visible color change from blue to green with a sensitivity of 20 and 0.15 ppb, respectively. In contrast, the dimethylamino-aza-BODIPY derivative, 3c, showed negligible affinity for the anions and neutral molecules tested. By tuning the photophysical properties through the judicious functionalization, the aza-BODIPY dyes thus synthesized can be utilized for the sensitive on-site detection and analysis of H2S, NO2(-), and NO in the aqueous medium.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium sulfide hydrate, ≥60%, scales
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium sulfide
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Nitromethane, puriss., absolute, over molecular sieve, ≥98.5% (GC)