Skip to Content
Merck
All Photos(3)

Key Documents

A7330

Sigma-Aldrich

Ammonium acetate

Ammonium acetate
1 of 1 reviewers received a sample product or took part in a promotion

BioXtra, ≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$42.00
500 G
$128.00
2.5 KG
$466.00

$42.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$42.00
500 G
$128.00
2.5 KG
$466.00

About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.25

$42.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

product line

BioXtra

Quality Level

Assay

≥98%

form

solid

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.01%

mp

110-112 °C (dec.) (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.001%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

N.CC(O)=O

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium acetate has been used:
  • to prepare ammonium acetate buffer[1]
  • in the preparation of mobile phase for LC/MS analysis[2]
  • as a supplement in establishing follicle culture[3]

Biochem/physiol Actions

Ammonium acetate can be used in proteinaceous baits and in beer waste to increase their attraction to Ceratitis capitate.[4] It helps in the progression of acetone-butanol-ethanol (ABE) fermentation when it is used as the only nitrogen source.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium acetate enhances the attractiveness of a variety of protein-based baits to female Ceratitis capitata (Diptera: Tephritidae)
Pinero JC, et al.
Journal of Economic Entomology, 108(2), 694-700 (2015)
DESI-MS as a tool for direct lipid analysis in cultured cells
Bodzon KA, et al.
Cytotechnology, 67(6), 1085-1091 (2015)
LC/MS/MS analysis of chloramphenicol in shrimp
Neuhaus BK, et al.
Laboratory Information Bulletin, 4290, 1-13 (2002)
Ammonium accumulation and use of mineral oil overlay do not alter imprinting establishment at three key imprinted genes in mouse oocytes grown and matured in a long-term follicle culture
Anckaert E, et al.
Biology of Reproduction, 81(4), 666-673 (2009)
Dual function of ammonium acetate in acetone-butanol-ethanol fermentation by Clostridium acetobutylicum
Li S, et al.
Bioresource Technology, 267, 319-325 (2018)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service