Skip to Content
Merck
All Photos(1)

Key Documents

71290

Sigma-Aldrich

Sodium azide

purum p.a., ≥99.0% (T)

Synonym(s):

Hydrazoic acid sodium salt, NSC 3072

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN3
CAS Number:
Molecular Weight:
65.01
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
form:
crystals
solubility:
H2O: 0.1 g/mL, clear, colorless

grade

purum p.a.

Quality Level

Assay

≥99.0% (T)

form

crystals

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na]N=[N+]=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Oxidative decarboxylation
  • Michael addition reactions

Reagent for synthesis of
  • Blue fluorescent copolymers
  • Metal phosphonates
  • Arenes via aminations

Involved in regioselective synthesis of prianosin B

Other Notes

Schmidt reaction with aldehydes and carboxylic acids, a review

suggested gloves for splash protection

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.I. Koldobski et al.
Russ. Chem. Rev., 47, 1084-1084 (1978)
V. N. Telvekar and K. A. Sasane,
Synlett, 18, 2778-2780 (2010)
Deng-Ke Cao et al.
Inorganic chemistry, 44(10), 3599-3604 (2005-05-10)
Based on the [hydroxy(4-pyridyl)methyl]phosphonate ligand, three compounds with formula Ni{(4-C(5)H(4)N)CH(OH)PO(3)}(H(2)O) (1), Cd{(4-C(5)H(4)N)CH(OH)PO(3)}(H(2)O) (2), and Gd{(4-C(5)H(4)N)CH(OH)P(OH)O(2)}(3).6H(2)O (3) have been synthesized under hydrothermal conditions. The crystal data for 1 are as follows: orthorhombic, space group Pbca, a = 8.7980(13) A, b =
H. Chen, et al.,
Macromolecules, 43, 3613-3623 (2010)
Y. Wada, et al.,
Tetrahedron, 65, 1059-1062 (2009)

Articles

Separation and identification of oligonucleotides differing by one base in length using Size Exclusion Chromatography combined with multi-angle light scattering (SEC-MALS) detector.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service