Skip to Content
Merck
All Photos(4)

Documents

372331

Sigma-Aldrich

Ammonium acetate

99.999% trace metals basis

Synonym(s):

Ammonium ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

crystalline
solid

technique(s)

HPLC: suitable

impurities

≤20.0 ppm Trace Metal Analysis

mp

110-112 °C (dec.) (lit.)

SMILES string

N.CC(O)=O

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium acetate is a white crystalline water-soluble ammonium salt with unique properties that make it suitable for use in solar cells, batteries, and fuel cell catalysts.

Application

Ammonium acetate can be used:
  • To passivate CsPbI3 perovskite nanocrystals for red LEDs. Ammonium acetate effectively eliminate undesired surface metallic lead cations and dangling bonds to enhance the stability and photoluminescence quantum yield (PLQY).
  • As an additive to fabricate high quality perovskite solar cell with enhanced power conversion efficiency (PCE) of 13.9%. It promotes crystallization and improve performance without changing the perovskite composition.
  • As a cost effective and green electrolyte for all-organic battery-supercapacitor hybrid device with maximum energy density.
  • As an electrolyte additive for aqueous zinc batteries. It helps to improve cycling stability and capacity retention.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anima Ghosal et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(3), 314-320 (2004-02-24)
Ezetimibe [1-(4-fluorophenyl)-3(R)-[3-(4-fluorophenyl)-3(S)-hydroxypropyl]-4(S)-(4-hydroxyphenyl)-2-azetidinone] (Zetia; Schering-Plough, Kenilworth, NJ) is the first in a new class of cholesterol-lowering agents known as cholesterol absorption inhibitors. The objective of this study was to identify the isoform(s) of human liver and intestinal UDP-glucuronosyltransferase (UGT) enzymes responsible
Reactions of phenanthraquinone and retenequinone with aldehydes and ammonium acetate in acetic acid solution1.
Steck EA and Day AR.
Journal of the American Chemical Society, 65(3), 452-456 (1943)
Selective inhibition of benzyl ether hydrogenolysis with Pd/C due to the presence of ammonia, pyridine or ammonium acetate.
Sajiki H.
Tetrahedron Letters, 36(20), 3465-3468 (1995)
Shi Hu et al.
Oncotarget, 6(3), 1695-1706 (2015-01-31)
Human epidermal growth factor receptors (HERs or ErbBs) play crucial roles in numerous cellular processes. ErbB2 is a key member of ErbB family, and its overexpression is recognized as a frequent molecular abnormality. In cancer, this overexpression correlates with aggressive
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service