Merck
  • Octacalcium phosphate ceramics combined with gingiva-derived stromal cells for engineered functional bone grafts.
Octacalcium phosphate ceramics combined with gingiva-derived stromal cells for engineered functional bone grafts.
Biomedical materials (Bristol, England) (2014-08-29)
Vadim L Zorin, Vladimir S Komlev, Alla I Zorina, Natalia V Khromova, Elena V Solovieva, Alexander Yu Fedotov, Ilya I Eremin, Pavel B Kopnin
ABSTRACT
Biocompatible ceramic fillers are capable of sustaining bone formation in the proper environment. The major drawback of these scaffolding materials is the absence of osteoinductivity. To overcome this limitation, bioengineered scaffolds combine osteoconductive components (biomaterials) with osteogenic features such as cells and growth factors. The bone marrow mesenchymal stromal cells (BMMSCs) and the β-tricalcium phosphate (β-TCP) are well-known and characterized in this regard. The present study was conducted to compare the properties of novel octacalcium phosphate ceramic (OCP) granules with β-TCP (Cerasorb(®)), gingiva-derived mesenchymal stromal cells (GMSCs) properties with the BMMSCs and osteogenic and angiogenic properties of a bioengineered composite based on OCP granules and the GMSCs. This study demonstrates that GMSCs and BMMSСs have a similar osteogenic capacity. The usage of OCP ceramic granules in combination with BMMSCs/GMSCs significantly affects the osteo- and angiogenesis in bone grafts of ectopic models.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
3,5,6-Trichloro-2-pyridinol, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium carbonate, ACS reagent, ≥30.0% NH3 basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium carbonate, puriss., meets analytical specification of NF, Ph. Franc., FCC
Proline, European Pharmacopoeia (EP) Reference Standard
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium carbonate, 99.999% trace metals basis
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard