Skip to Content
Merck
All Photos(4)

Key Documents

900503

Sigma-Aldrich

Sodium carbonate

anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$129.00
1 KG
$182.00
2.5 KG
$251.00
5 KG
$453.00
12 KG
$741.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$129.00
1 KG
$182.00
2.5 KG
$251.00
5 KG
$453.00
12 KG
$741.00

About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder or granules

$129.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for EPA 300

product line

Redi-Dri

Assay

≥99.5%

form

powder or granules

quality

free-flowing

impurities

≤0.005% silica
≤0.01% insolubles

loss

≤1.0% loss on heating, 285°C

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.03%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angeles M Alonso et al.
Journal of agricultural and food chemistry, 50(21), 5832-5836 (2002-10-03)
It has been demonstrated that wine and other products derived from the grape have a high antioxidant capability; as a possible consequence of this, they may have potential benefits for health. The byproducts of the winemaking process represent a source
Mark Wahrenbrock et al.
The Journal of clinical investigation, 112(6), 853-862 (2003-09-17)
Trousseau described spontaneous, recurrent superficial migratory thrombophlebitis associated with occult cancers, and this was later correlated with disseminated microangiopathy (platelet-rich clots in small blood vessels). Trousseau syndrome often occurs with mucinous adenocarcinomas, which secrete abnormally glycosylated mucins and mucin fragments
Jian-Wen Chen et al.
American journal of physiology. Endocrinology and metabolism, 284(6), E1149-E1155 (2003-02-27)
At present, the circulating bioactivity of insulin-like growth factor I (IGF-I) is estimated by immunological measurements of IGF-I levels. However, immunoassays ignore the modifying effects of the IGF-binding proteins (IGFBPs) on the interaction between IGF-I and the IGF-I receptor (IGF-IR).
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)
Yukiyo Sato et al.
PloS one, 9(6), e99041-e99041 (2014-06-11)
The accumulation of RCY1 protein, which is encoded by RESISTANCE TO CMV(Y) (RCY1), a CC-NB-LRR class R-gene, is tightly correlated with the strength of the resistance to a yellow strain of Cucumber mosaic virus [CMV(Y)] in Arabidopsis thaliana. In order

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service