Skip to Content
Merck
All Photos(3)

Key Documents

223530

Sigma-Aldrich

Sodium carbonate

Sodium carbonate

powder, ≥99.5%, ACS reagent

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$260.00
1 KG
$371.00
2.5 KG
$655.00
12 KG
$1,490.00

$260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.

Request a Bulk Order

Select a Size

Change View
500 G
$260.00
1 KG
$371.00
2.5 KG
$655.00
12 KG
$1,490.00

About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder

$260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability
This SKU is also available as ACS grade in our Exclusive Redi-Dri™ free-flowing product line.

Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

suitable for EPA 300
suitable for SM 2310
suitable for SM 2320
suitable for SM 4500 - NH3

Assay

≥99.5%

form

powder

impurities

≤0.005% silica
≤0.01% insolubles

loss

≤1.0% loss on heating

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.03%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate (soda ash) is an inorganic chemical compound. It is used in the synthesis of chemicals, detergent formulations, flue gas desulfurization, and paper and pulp production.

Application

Sodium carbonate can be used:
  • As a catalyst in the aldol condensation of aldehydes with ketones in the synthesis of chalcone and azachalcone.
  • As a pH stabilizer agent in the synthesis of FeO-NPs (iron oxide nanoparticles) in the presence of cymbopogon citratus as a reducing agent.
  • As a base in the Pd-catalyzed Suzuki coupling reactions.
  • As a base in the transition metal free Suzuki coupling reaction of aryl halides and arylboronic acids to form biaryls using water as a solvent and TBAB (tetrabutylammonium bromide) as a phase-transfer catalyst.

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angeles M Alonso et al.
Journal of agricultural and food chemistry, 50(21), 5832-5836 (2002-10-03)
It has been demonstrated that wine and other products derived from the grape have a high antioxidant capability; as a possible consequence of this, they may have potential benefits for health. The byproducts of the winemaking process represent a source
Jian-Wen Chen et al.
American journal of physiology. Endocrinology and metabolism, 284(6), E1149-E1155 (2003-02-27)
At present, the circulating bioactivity of insulin-like growth factor I (IGF-I) is estimated by immunological measurements of IGF-I levels. However, immunoassays ignore the modifying effects of the IGF-binding proteins (IGFBPs) on the interaction between IGF-I and the IGF-I receptor (IGF-IR).
Mark Wahrenbrock et al.
The Journal of clinical investigation, 112(6), 853-862 (2003-09-17)
Trousseau described spontaneous, recurrent superficial migratory thrombophlebitis associated with occult cancers, and this was later correlated with disseminated microangiopathy (platelet-rich clots in small blood vessels). Trousseau syndrome often occurs with mucinous adenocarcinomas, which secrete abnormally glycosylated mucins and mucin fragments
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)
Yukiyo Sato et al.
PloS one, 9(6), e99041-e99041 (2014-06-11)
The accumulation of RCY1 protein, which is encoded by RESISTANCE TO CMV(Y) (RCY1), a CC-NB-LRR class R-gene, is tightly correlated with the strength of the resistance to a yellow strain of Cucumber mosaic virus [CMV(Y)] in Arabidopsis thaliana. In order

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service