Skip to Content
Merck
All Photos(1)

Key Documents

PHR1247

Supelco

Indomethacin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

IDM, 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$149.00

$149.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$149.00

About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
Beilstein:
497341
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$149.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. I0200000
traceable to USP 1341001

API family

indomethacin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1ccc2n(c(C)c(CC(O)=O)c2c1)C(=O)c3ccc(Cl)cc3

InChI

1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)

InChI key

CGIGDMFJXJATDK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Indomethacin drug is extensively employed as a pharmacologic agent to bring about the closure of patent ductus arteriosus (PDA) in premature infants.[1]

Application

Indomethacin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and plasma samples by chromatography techniques.[2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that is relatively selective for COX-1.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0281 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Liquid chromatographic determination of atorvastatin in bulk drug, tablets, and human plasma
Altuntas TG and Erk N
Journal of Liquid Chromatography and Related Technologies, 27(1), 83-93 (2004)
Simple, rapid and sensitive method for the determination of indomethacin in plasma by high-performance liquid chromatography with ultraviolet detection
Sato J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 692(1), 241-244 (1997)
Advantages of application of UPLC in pharmaceutical analysis.
Novakova L, et al.
Talanta, 68(3), 908-918 (2006)
Jason P Riley et al.
Clinical science (London, England : 1979), 125(11), 533-542 (2013-06-27)
One feature of allergic asthma, the EAR (early allergic reaction), is not present in the commonly used mouse models. We therefore investigated the mediators involved in EAR in a guinea-pig in vivo model of allergic airway inflammation. Animals were sensitized
Dongfei Liu et al.
Journal of controlled release : official journal of the Controlled Release Society, 170(2), 268-278 (2013-06-13)
Nanoparticulate drug delivery systems offer remarkable opportunities for clinical treatment. However, there are several challenges when they are employed to deliver multiple cargos/payloads, particularly concerning the synchronous delivery of small molecular weight drugs and relatively larger peptides. Since porous silicon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service