Merck
  • Simultaneous quantification of lenalidomide, ibrutinib and its active metabolite PCI-45227 in rat plasma by LC-MS/MS: application to a pharmacokinetic study.
Simultaneous quantification of lenalidomide, ibrutinib and its active metabolite PCI-45227 in rat plasma by LC-MS/MS: application to a pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2015-01-17)
Sridhar Veeraraghavan, Srikant Viswanadha, Satheeshmanikandan Thappali, Babu Govindarajulu, Swaroopkumar Vakkalanka, Manivannan Rangasamy
ABSTRACT
Efficacy assessments using a combination of ibrutinib and lenalidomide necessitate the development of an analytical method for determination of both drugs in plasma with precision. A high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed for the simultaneous determination of lenalidomide, ibrutinib, and its active metabolite PCI45227 in rat plasma. Extraction of lenalidomide, ibrutinib, PCI45227 and tolbutamide (internal standard; IS) from 50 μl rat plasma was carried out by liquid-liquid extraction with ethyl acetate:dichloromethane (90:10) ratio. Chromatographic separation of analytes was performed on YMC pack ODS AM (150 mm × 4.6 mm, 5 μm) column under gradient conditions with acetonitrile:0.1% formic acid buffer as the mobile phases at a flow rate of 1 ml/min. Precursor ion and product ion transition for analytes and IS were monitored on a triple quadrupole mass spectrometer, operated in the selective reaction monitoring with positive ionization mode. Method was validated over a concentration range of 0.72-183.20 ng/ml for ibrutinib, 0.76-194.33 ng/ml for PCI-45227 and 1.87-479.16 ng/ml for lenalidomide. Mean extraction recovery for ibrutinib, PCI-45227, lenalidomide and IS of 75.2%, 84.5%, 97.3% and 92.3% were consistent across low, medium, and high QC levels. Precision and accuracy at low, medium and high quality control levels were less than 15% across analytes. Bench top, wet, freeze-thaw and long term stability was evaluated for all the analytes. The analytical method was applied to support a pharmacokinetic study of simultaneous estimation of lenalidomide, ibrutinib, and its active metabolite PCI-45227 in Wistar rat. Assay reproducibility was demonstrated by re-analysis of 18 incurred samples.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Supelco
Tolbutamide, analytical standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available. Contact Technical Service
Supelco
Ethyl acetate, analytical standard
Pricing and availability is not currently available. Contact Technical Service
Supelco
Dichloromethane, Selectophore, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Pricing and availability is not currently available. Contact Technical Service
Supelco
Dichloromethane, analytical standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available. Contact Technical Service
Tolbutamide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Pricing and availability is not currently available. Contact Technical Service