Merck
  • Sources and fate of perfluorinated compounds in the aqueous environment and in drinking water of a highly urbanized and industrialized area in Italy.
Sources and fate of perfluorinated compounds in the aqueous environment and in drinking water of a highly urbanized and industrialized area in Italy.
Journal of hazardous materials (2014-07-06)
Sara Castiglioni, Sara Valsecchi, Stefano Polesello, Marianna Rusconi, Manuela Melis, Marinella Palmiotto, Angela Manenti, Enrico Davoli, Ettore Zuccato
ABSTRACT
Perfluorinated substances are listed among emerging contaminants because they are globally distributed, environmentally persistent, bioaccumulative and potentially harmful. In a three-year monitoring campaign (2010-2013) we investigated the occurrence, sources and fate of nine perfluoroalkylcarboxylic acids and three perfluoroalkylsulfonic acids, in the most industrialized region of Italy. Composite samples were collected in influents and effluents of wastewater treatment plants (WWTPs), in the main rivers flowing through the basin, and in raw groundwater and finished drinking water. Samples were analyzed by liquid chromatography tandem mass spectrometry. Perfluorinated substances were not removed in WWTPs and those receiving industrial wastes discharged up to 50 times the loads of WWTPs receiving municipal wastes. The mass balance of the emissions in the River Lambro basin showed continuously increasing contamination from north to south and differences in the composition of homologues in the west and east sides of the basin. Ground and drinking water were contaminated in industrial areas, but these substances were removed well in Milan. Contamination from industrial sources was prevalent over urban sources, contributing to 90% of the loads measured at the closure of the basin. The River Lambro was confirmed as one of the main sources of contamination in the Po River.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Methanol, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%