Skip to Content
Merck
All Photos(2)

Key Documents

171468

Sigma-Aldrich

Perfluorooctanoic acid

95%

Synonym(s):

Pentadecafluorooctanoic acid, PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$56.40
25 G
$187.00

$56.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$56.40
25 G
$187.00

About This Item

Linear Formula:
CF3(CF2)6COOH
CAS Number:
Molecular Weight:
414.07
Beilstein:
1809678
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.22

$56.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

bp

189 °C/736 mmHg (lit.)

mp

55-56 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8HF15O2/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23/h(H,24,25)

InChI key

SNGREZUHAYWORS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

PFOA, its salts and related substances are listed in Part A of Annex I of the Persistent Organic Pollutants (POPs) Regulation (EU) 2019/1021. It can only be used for Laboratory-scale research or as a reference standard

Derogations until 4 July 2025 are uses in
  • photolithography or etch processes in semiconductor manufacturing
  • photographic coatings applied to films
  • invasive and implantable medical devices
The use is further allowed in firefighting foams for Class B fires until 4 July 2025

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - Skin Corr. 1C - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding of perfluorooctanoic acid to rat and human plasma proteins.
Han X, et al.
Chemical Research in Toxicology, 16(6), 775-781 (2003)
Perfluoroalkyl acids: a review of monitoring and toxicological findings
C Lau, et al.
Toxicological Sciences, 99, 366-394 (2007)
Effects of perfluorooctanoic acid exposure during pregnancy in the mouse.
Lau C, et al.
Toxicological Sciences, 90(2), 510-518 (2006)
Lilan Zhang et al.
Environmental pollution (Barking, Essex : 1987), 229, 159-167 (2017-06-09)
With the phaseout of perfluorooctane sulfonate (PFOS) production in most countries and its well known recalcitrance, there is a need to quantify the potential release of PFOS from precursors previously or currently being emitted into the environment. Aerobic biodegradation of
Polyfluorinated compounds: past, present, and future
AB Lindstrom, et al.
Environmental Science & Technology, 45, 7954-7961 (2011)

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the solubility of Heptadecafluorooctanesulfonic acid potassium salt product 77282 (CAS 2795-39-3)?

    1 answer
    1. The solubility for 77282 has not been validated, but the product should be soluble in methanol and slightly soluble (miscible) in water (with heating and sonication).

      Helpful?

  4. What is the solubility of Perfluorononanoic acid (product 394459, CAS 375-95-1)?

    1 answer
    1. The solubility of the product has not been validated, however, it is expected to be soluble in DMSO, methanol, and acetone, and slightly soluble in water.

      Helpful?

  5. What is the solubility of Perfluorooctanoic acid (171468, CAS 335-67-1)?

    1 answer
    1. The solubility for this item has not been validated, but the product should be readily soluble in water. It may also be soluble in chloroform and methanol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service