Merck
  • Evaluation of the sensitivity, responsiveness and reproducibility of primary rainbow trout hepatocyte vitellogenin expression as a screening assay for estrogen mimics.
Evaluation of the sensitivity, responsiveness and reproducibility of primary rainbow trout hepatocyte vitellogenin expression as a screening assay for estrogen mimics.
Aquatic toxicology (Amsterdam, Netherlands) (2015-01-07)
Maria T Hultman, Jan Thomas Rundberget, Knut Erik Tollefsen
ABSTRACT
Vitellogenin (Vtg) induction in primary fish hepatocytes has been proposed as an in vitro screening assay for ER agonists and antagonists, but has not yet been used extensively as a high-throughput screening tool due to poor reproducibility, sensitivity and overall feasibility. The present work has evaluated the role of seasonal variation, normalization, optimal culture and assay conditions on the sensitivity, responsiveness and reproducibility of in vitro vtg gene mRNA and protein expression in rainbow trout (Oncorhynchus mykiss) primary hepatocytes using the xenoestrogen 17α-ethynylestradiol (EE2) as a test chemical. The results show that primary hepatocytes display a relatively high individual and seasonal variation in both Vtg mRNA and protein induction potential, although less variance was observed in assay sensitivity. Data normalization of assay response to maximum (3 nM EE2) and minimum (DMSO) Vtg production dramatically reduced this variance and led to improved assay reproducibility. A time-dependent response was observed both for mRNA and protein expression, reaching maximum Vtg induction after 96 h of exposure, although reproducible concentration response curves for both Vtg mRNA and protein could be obtained already after 48 h. A need for chemical re-exposure of the hepatocytes was identified to be important for sustaining exposure concentrations in extended studies (>48 h), whereas different plate formats (96, 24 or 6 wells) did not affect the bioassay performance. In conclusion, standardization of hepatocyte bioassay and test conditions as well as data-normalization procedures are proposed to be instrumental for more consistent and comparable results in future use of this in vitro assay.
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Acetone, natural, ≥97%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Pricing and availability is not currently available. Contact Technical Service
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Supelco
Acetone, analytical standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Pricing and availability is not currently available. Contact Technical Service
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Pricing and availability is not currently available. Contact Technical Service
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service