Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Pricing and availability is not currently available.